Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT61B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686858.3 | XP_005686915.1 | 4 - 5 | 1769 | 82 | |
XM_005686859.3 | XP_005686916.1 | 4 - 5 | 1768 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |