Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058978.1 | XP_017914467.1 | 1 - 2 | 171 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |