Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18H16orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692120.3 | XP_005692177.1 | 9 - 10 | 798 | 78 | |
XM_005692121.3 | XP_005692178.1 | 9 - 10 | 784 | 78 | ||
XM_018062103.1 | XP_017917592.1 | 9 - 10 | 786 | 78 | ||
GenBank mRNA | JO587830.2 | - | 2 - 3 | 151 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |