Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058697.1 | XP_017914186.1 | 1 - 2 | 409 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |