Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689731.3 | XP_005689788.2 | 6 - 7 | 2524 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |