Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693262.3 | XP_005693319.1 | 1 - 2 | 471 | 99 | |
GenBank mRNA | JT675845.1 | - | 3 - 4 | 1631 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |