Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049045.1 | XP_017904534.1 | 5 - 6 | 690 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |