Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059371.1 | XP_017914860.1 | 3 - 4 | 310 | 102 | |
GenBank mRNA | JO601901.2 | - | 3 - 4 | 318 | 102 | |
JO636706.2 | - | 3 - 4 | 369 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |