Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059035.1 | XP_017914524.1 | 10 - 11 | 1201 | 64 | |
GenBank mRNA | JO430053.2 | - | 10 - 11 | 1012 | 64 | |
JO632649.2 | - | 10 - 11 | 1355 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |