Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675884.3 | XP_005675941.1 | 1 - 2 | 399 | 79 | |
GenBank mRNA | KA335585.1 | - | 1 - 2 | 361 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |