Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684767.3 | XP_005684824.2 | 6 - 7 | 1377 | 94 | |
XM_005684768.3 | XP_005684825.2 | 6 - 7 | 1377 | 94 | ||
GenBank mRNA | JO421239.2 | - | 6 - 7 | 1400 | 94 | |
JO430014.2 | - | 2 - 3 | 289 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |