Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053770.1 | XP_017909259.1 | 3 - 4 | 764 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |