Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JT679346.1 | - | 6 - 7 | 543 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |