Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013969300.2 | XP_013824754.2 | 5 - 6 | 2929 | 115 | |
GenBank mRNA | JT683129.1 | - | 5 - 6 | 1846 | 115 | |
KA316736.1 | - | 1 - 2 | 471 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |