Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692751.1 | XP_005692808.1 | 3 - 4 | 316 | 91 | |
GenBank mRNA | KA340850.1 | - | 3 - 4 | 305 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |