Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042602.1 | XP_017898091.1 | 3 - 4 | 544 | 81 | |
GenBank mRNA | JO637798.2 | - | 1 - 2 | 285 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |