Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677142.3 | XP_005677199.3 | 3 - 4 | 677 | 107 | |
GenBank mRNA | JO503947.2 | - | 3 - 4 | 688 | 107 | |
JO643382.2 | - | 3 - 4 | 638 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |