Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061864.1 | XP_017917353.1 | 4 - 5 | 446 | 82 | |
XM_018061865.1 | XP_017917354.1 | 4 - 5 | 387 | 82 | ||
GenBank mRNA | JO578964.1 | - | 4 - 5 | 491 | 82 | |
JO643282.2 | - | 4 - 5 | 590 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |