Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060657.1 | XP_017916146.1 | 7 - 8 | 950 | 61 | |
GenBank mRNA | JO431307.2 | - | 1 - 2 | 901 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |