Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062404.1 | XP_017917893.1 | 7 - 8 | 3417 | 62 | |
XM_018062405.1 | XP_017917894.1 | 6 - 7 | 2169 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |