Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043250.1 | XP_017898739.1 | 6 - 7 | 1533 | 84 | |
XM_018043251.1 | XP_017898740.1 | 6 - 7 | 1449 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |