Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3H1orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678598.3 | XP_005678655.3 | 2 - 3 | 247 | 59 | |
XM_018042680.1 | XP_017898169.1 | 2 - 3 | 238 | 59 | ||
GenBank mRNA | JO438657.2 | - | 2 - 3 | 227 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |