Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102177559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692520.2 | XP_005692577.2 | 1 - 2 | 78 | 90 | |
XM_013971097.2 | XP_013826551.2 | 1 - 2 | 78 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |