Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051147.1 | XP_017906636.1 | 2 - 3 | 309 | 68 | |
XM_018051148.1 | XP_017906637.1 | 2 - 3 | 1337 | 68 | ||
XM_018051149.1 | XP_017906638.1 | 2 - 3 | 316 | 68 | ||
GenBank mRNA | JO439178.2 | - | 2 - 3 | 281 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |