Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053260.1 | XP_017908749.1 | 3 - 4 | 727 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |