Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044964.1 | XP_017900453.1 | 7 - 8 | 1015 | 88 | |
XM_018044965.1 | XP_017900454.1 | 7 - 8 | 994 | 88 | ||
GenBank mRNA | JO433756.2 | - | 7 - 8 | 1067 | 88 | |
KA341255.1 | - | 1 - 2 | 248 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |