Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057390.1 | XP_017912879.1 | 4 - 5 | 369 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |