Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689523.3 | XP_005689580.2 | 3 - 4 | 375 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |