Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TATDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038323.1 | XP_017893812.1 | 6 - 7 | 2733 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |