Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699180.2 | XP_005699237.1 | 3 - 4 | 774 | 58 | |
GenBank mRNA | JP351470.2 | - | 6 - 7 | 1528 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |