Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAC3D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699983.3 | XP_005700040.2 | 2 - 3 | 730 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |