Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049263.1 | XP_017904752.1 | 2 - 3 | 623 | 100 | |
GenBank mRNA | JO618711.2 | - | 1 - 2 | 192 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |