Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685207.3 | XP_005685264.1 | 3 - 4 | 1104 | 81 | |
XM_018054643.1 | XP_017910132.1 | 3 - 4 | 411 | 81 | ||
XM_018054644.1 | XP_017910133.1 | 3 - 4 | 401 | 81 | ||
GenBank mRNA | JO437600.2 | - | 2 - 3 | 776 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |