Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044973.1 | XP_017900462.1 | 6 - 7 | 754 | 187 | |
GenBank mRNA | JO424441.1 | - | 6 - 7 | 831 | 187 | |
KA330779.1 | - | 4 - 5 | 1448 | 187 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |