Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677230.2 | XP_005677287.1 | 1 - 2 | 601 | 65 | |
GenBank mRNA | JO536690.2 | - | 1 - 2 | 643 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |