Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682378.2 | XP_005682435.1 | 1 - 2 | 76 | 118 | |
XM_018051744.1 | XP_017907233.1 | 1 - 2 | 76 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |