Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042167.1 | XP_017897656.1 | 5 - 6 | 1035 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |