Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051608.1 | XP_017907097.1 | 4 - 5 | 338 | 91 | |
XM_018051609.1 | XP_017907098.1 | 4 - 5 | 333 | 91 | ||
GenBank mRNA | JT618983.1 | - | 4 - 5 | 287 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Amplicon length between 86 and 100 Capra hircus Probe spans exons |