Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063837.1 | XP_017919326.1 | 1 - 2 | 3045 | 88 | |
GenBank mRNA | JO617695.2 | - | 1 - 2 | 345 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |