Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675457.3 | XP_005675514.1 | 4 - 5 | 513 | 67 | |
XM_013965820.2 | XP_013821274.1 | 3 - 4 | 509 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |