Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051186.1 | XP_017906675.1 | 2 - 3 | 723 | 104 | |
XM_018051192.1 | XP_017906681.1 | 2 - 3 | 723 | 104 | ||
XM_018051198.1 | XP_017906687.1 | 2 - 3 | 723 | 104 | ||
XM_018051204.1 | XP_017906693.1 | 2 - 3 | 723 | 104 | ||
XM_018051213.1 | XP_017906702.1 | 2 - 3 | 349 | 104 | ||
XM_018051219.1 | XP_017906708.1 | 2 - 3 | 342 | 104 | ||
GenBank mRNA | JO581787.1 | - | 3 - 4 | 1441 | 104 | |
JO581788.1 | - | 3 - 4 | 984 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |