Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHEJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676555.3 | XP_005676612.2 | 3 - 4 | 728 | 67 | |
XM_018059284.1 | XP_017914773.1 | 3 - 4 | 633 | 67 | ||
GenBank mRNA | JT671972.1 | - | 7 - 8 | 1021 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |