Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053417.1 | XP_017908906.1 | 3 - 4 | 256 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |