Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041753.1 | XP_017897242.1 | 1 - 2 | 566 | 86 | |
GenBank mRNA | JT691795.1 | - | 1 - 2 | 415 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |