Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699311.3 | XP_005699368.1 | 16 - 17 | 3798 | 75 | |
XM_018042447.1 | XP_017897936.1 | 16 - 17 | 3785 | 75 | ||
GenBank mRNA | JI861778.1 | - | 14 - 15 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |