Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040795.1 | XP_017896284.1 | 1 - 2 | 246 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |