Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047040.1 | XP_017902529.1 | 3 - 4 | 6724 | 97 | |
XM_018047042.1 | XP_017902531.1 | 4 - 5 | 5873 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |