Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MC3R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688325.2 | XP_005688382.1 | 1 | 421 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |