Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057604.1 | XP_017913093.1 | 4 - 5 | 979 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |