Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013970630.2 | XP_013826084.1 | 7 - 8 | 942 | 102 | |
XM_013970631.2 | XP_013826085.1 | 7 - 8 | 1014 | 102 | ||
XM_013970632.2 | XP_013826086.1 | 7 - 8 | 880 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |